Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009155.hg.1 |
ENST00000623103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009155.hg.1 |
ENST00000623103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009155.hg.1 |
ENST00000623103.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009155.hg.1 |
NM_173520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009155.hg.1 |
NM_173520.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009155.hg.1 |
NM_173520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162333.hg.1 |
ENST00000623103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162333.hg.1 |
NM_173520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162333.hg.1 |
ENST00000623103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162333.hg.1 |
NM_173520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162333.hg.1 |
TC0900009155.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900009155.hg.1 |
ENST00000623103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009155.hg.1 |
ENST00000623103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009155.hg.1 |
ENST00000623103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009155.hg.1 |
NM_173520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009155.hg.1 |
NM_173520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009155.hg.1 |
NM_173520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162334.hg.1 |
ENST00000623103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162334.hg.1 |
NM_173520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162334.hg.1 |
ENST00000623103.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900162334.hg.1 |
NM_173520.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900162334.hg.1 |
TC0900009155.hg.1 |
Clariom D TC |