Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000531711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000531711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000531711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000531711.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000541883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
ENST00000541883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000420873.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000434798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000531711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000531711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000531711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000531711.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000541883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
ENST00000541883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006611.hg.1 |
NR_027138.1 |
Refseq |