Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173653.3 | NP_775924.1 | 2 - 3 | 588 | 90 | |
XM_011512704.2 | XP_011511006.1 | 2 - 3 | 561 | 90 | ||
XM_017006202.1 | XP_016861691.1 | 2 - 3 | 561 | 90 | ||
GenBank mRNA | AB089794.1 | - | 2 - 3 | 379 | 90 | |
AK315107.1 | - | 2 - 3 | 515 | 90 | ||
AL832304.1 | - | 2 - 3 | 453 | 90 | ||
AY254100.1 | - | 2 - 3 | 503 | 90 | ||
BC035779.1 | - | 2 - 3 | 504 | 5590356 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons |