Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022791.hg.1 |
TC09001763.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022791.hg.1 |
ENST00000321773.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022791.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022791.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186989.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186989.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186989.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186989.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186989.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186989.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186989.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186989.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186989.hg.1 |
TC0900012294.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000333046.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000409012.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
ENST00000477345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012294.hg.1 |
NM_001128228.2 |
Refseq |