Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280096.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000480534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000480534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000480534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000485653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000485653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000485653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000485653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
ENST00000485653.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024074.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013220.hg.1 |
TC02000891.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013220.hg.1 |
ENST00000280097.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013220.hg.1 |
ENST00000329366.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013220.hg.1 |
ENST00000410115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013220.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013220.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000480534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
ENST00000480534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200166334.hg.1 |
TC0200009470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000280097.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000329366.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000410115.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000467390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000475675.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000480534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000480534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000480534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000485653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000485653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000485653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000485653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
ENST00000485653.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024074.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_001024075.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009470.hg.1 |
NM_006895.2 |
Refseq |