Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000306031.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000370192.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000460019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000460019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000460019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000460019.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000474241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000474241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000474241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000474241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000474241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
ENST00000474241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_000110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015023.hg.1 |
NM_001160301.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000629811.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000629811.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000630798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
ENST00000630798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000422980.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000629811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000629811.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000630798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
ENST00000630798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009153.hg.1 |
NR_046590.1 |
Refseq |