Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000580242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000580242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003225.hg.1 |
ENST00000400256.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003225.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003225.hg.1 |
TC18000273.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000580242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000580242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007850.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800153737.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800153737.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800153737.hg.1 |
NM_130386.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800153737.hg.1 |
TC1800007850.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800153737.hg.1 |
ENST00000400256.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800153737.hg.1 |
ENST00000582147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800153737.hg.1 |
NM_130386.2 |
Refseq |