Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010489.hg.1 |
TC01000641.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010489.hg.1 |
ENST00000257181.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010489.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010489.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159257.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159257.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159257.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159257.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159257.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100159257.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159257.hg.1 |
TC0100008315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000257181.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000474048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
ENST00000487160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008315.hg.1 |
NM_032864.3 |
Refseq |