Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000180.hg.1 |
TC16002048.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000180.hg.1 |
ENST00000219409.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000180.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011316.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
ENST00000219409.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
ENST00000414650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
ENST00000447871.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
ENST00000477621.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
NM_001176.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600188490.hg.1 |
TC1600011316.hg.1 |
Clariom D TC |