Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005347.4 | NP_005338.1 | 1 - 2 | 385 | 146 | |
GenBank mRNA | AF216292.1 | - | 1 - 2 | 124 | 146 | |
AJ271729.1 | - | 1 - 2 | 137 | 146 | ||
AK293604.1 | - | 1 - 2 | 341 | 146 | ||
BC020235.1 | - | 1 - 2 | 328 | 5020098 | 146 | |
EU794617.1 | - | 1 - 2 | 137 | 146 | ||
X87949.1 | - | 1 - 2 | 346 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons |