Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07020348.hg.1 |
TC07001319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07020348.hg.1 |
ENST00000395925.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07020348.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07020348.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000428534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000428534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000428534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000428534.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000437480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000437480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000437480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000437480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000437480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000464291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000464291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000464291.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07020349.hg.1 |
TC07001319.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07020349.hg.1 |
ENST00000395925.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07020349.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07020349.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177176.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177176.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177176.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177176.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177176.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177176.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177176.hg.1 |
TC0700010857.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000428534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000428534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000428534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000428534.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000437480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000437480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000437480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000437480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000437480.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000448703.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000464291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000464291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000464291.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010857.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177175.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177175.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177175.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177175.hg.1 |
ENST00000395925.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177175.hg.1 |
ENST00000479210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700177175.hg.1 |
NM_000168.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700177175.hg.1 |
TC0700010857.hg.1 |
Clariom D TC |