Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003744.hg.1 |
TC03000192.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003744.hg.1 |
ENST00000352511.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003744.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003743.hg.1 |
TC03000192.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003743.hg.1 |
ENST00000352511.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03003743.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149559.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149559.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149559.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149559.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149559.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149559.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149559.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149559.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149559.hg.1 |
TC0300007077.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007077.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149557.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149557.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149557.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149557.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149557.hg.1 |
ENST00000352511.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149557.hg.1 |
ENST00000461232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149557.hg.1 |
ENST00000465020.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300149557.hg.1 |
NM_001106.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300149557.hg.1 |
TC0300007077.hg.1 |
Clariom D TC |