Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028860.hg.1 |
TC06002304.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028860.hg.1 |
ENST00000296946.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028860.hg.1 |
ENST00000366871.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028860.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600195598.hg.1 |
TC0600013870.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000296946.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366871.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000366876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
ENST00000461348.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_001270484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013870.hg.1 |
NM_003181.3 |
Refseq |