Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004106.1 | NP_004097.1 | 3 - 4 | 201 | 59 | |
GenBank mRNA | AK307102.1 | - | 3 - 4 | 226 | 59 | |
BC033872.1 | - | 3 - 4 | 198 | 5219148 | 59 | |
M33195.1 | - | 3 - 4 | 201 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |