Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014243.2 | NP_055058.2 | 2 - 3 | 129 | 106 | |
GenBank mRNA | AB002364.1 | - | 2 - 3 | 81 | 106 | |
AF247668.1 | - | 2 - 3 | 129 | 106 | ||
BC130287.1 | - | 2 - 3 | 126 | 40146318 | 106 | |
BC132735.1 | - | 2 - 3 | 126 | 40146757 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons |