Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEL1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297592.1 | NP_001284521.1 | 14 - 15 | 2256 | 70 | |
NM_001297594.1 | NP_001284523.1 | 14 - 15 | 2278 | 70 | ||
NM_015187.4 | NP_056002.2 | 14 - 15 | 2473 | 70 | ||
XM_011513819.2 | XP_011512121.2 | 14 - 15 | 2467 | 70 | ||
XM_017007923.1 | XP_016863412.1 | 13 - 14 | 2111 | 70 | ||
GenBank mRNA | AB018289.1 | - | 13 - 14 | 1971 | 70 | |
AK025282.1 | - | 2 - 3 | 139 | 70 | ||
AK289687.1 | - | 14 - 15 | 2221 | 70 | ||
AK300219.1 | - | 5 - 6 | 571 | 70 | ||
BC009945.2 | - | 7 - 8 | 988 | 70 | ||
BC029422.1 | - | 4 - 5 | 398 | 70 | ||
BC041046.1 | - | 5 - 6 | 625 | 70 | ||
BC060867.1 | - | 14 - 15 | 2266 | 30344157 | 70 | |
CR749817.1 | - | 14 - 15 | 2256 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |