Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000491437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000491437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000491437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000491437.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000495916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000495916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
ENST00000495916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008474.hg.1 |
TC07000579.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008474.hg.1 |
ENST00000319273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008474.hg.1 |
ENST00000350485.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008474.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008474.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000346867.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000491437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000491437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000491437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000491437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000495916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000495916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
ENST00000495916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013997.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008435.hg.1 |
NM_013998.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
ENST00000491437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
ENST00000319273.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
ENST00000350485.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
ENST00000491437.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
NM_003182.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
NM_013996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158840.hg.1 |
TC0700008435.hg.1 |
Clariom D TC |