Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022873.hg.1 |
TC09001770.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022873.hg.1 |
ENST00000340951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022873.hg.1 |
ENST00000342129.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022873.hg.1 |
NM_017820.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022875.hg.1 |
TC09001770.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022875.hg.1 |
ENST00000340951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022875.hg.1 |
ENST00000342129.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022875.hg.1 |
NM_017820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
ENST00000340951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
ENST00000472958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
ENST00000490886.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
ENST00000491734.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
NM_017820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
ENST00000340951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
ENST00000472958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
ENST00000490886.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
ENST00000491734.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
NM_017820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183024.hg.1 |
TC0900012055.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
ENST00000340951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
ENST00000472958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
ENST00000478344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
ENST00000490886.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
ENST00000491734.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
NM_017820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
ENST00000340951.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
ENST00000472958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
ENST00000478344.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
ENST00000490886.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
ENST00000491734.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
NM_017820.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900183022.hg.1 |
TC0900012055.hg.1 |
Clariom D TC |