Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000461650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000461650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000461650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000461650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000468698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000468698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
ENST00000468698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006943.hg.1 |
TC10002941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006943.hg.1 |
ENST00000371924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006943.hg.1 |
ENST00000371926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006943.hg.1 |
ENST00000371927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006943.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006944.hg.1 |
TC10002941.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006944.hg.1 |
ENST00000371924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006944.hg.1 |
ENST00000371926.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006944.hg.1 |
ENST00000371927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10006944.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000461650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000461650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000461650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000461650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000468698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000468698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
ENST00000468698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008385.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156811.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156811.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156811.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156811.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156811.hg.1 |
ENST00000371924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156811.hg.1 |
ENST00000371926.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156811.hg.1 |
ENST00000371927.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000156811.hg.1 |
NM_020799.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000156811.hg.1 |
TC1000008385.hg.1 |
Clariom D TC |