Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022786.hg.1 |
TC01001465.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022786.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022785.hg.1 |
TC01001465.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022785.hg.1 |
ENST00000367833.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022785.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022784.hg.1 |
TC01001465.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022784.hg.1 |
ENST00000367833.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022784.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177475.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177475.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177475.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177475.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177475.hg.1 |
TC0100010522.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100177476.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177476.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177476.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177476.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177476.hg.1 |
TC0100010522.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100177474.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177474.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177474.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177474.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177474.hg.1 |
TC0100010522.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_001031800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010522.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177477.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177477.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177477.hg.1 |
ENST00000367833.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100177477.hg.1 |
NM_152902.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100177477.hg.1 |
TC0100010522.hg.1 |
Clariom D TC |