Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004335.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004335.hg.1 |
ENST00000545142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004335.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004335.hg.1 |
TC11000342.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004334.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004334.hg.1 |
ENST00000524419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004334.hg.1 |
ENST00000528989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004334.hg.1 |
ENST00000545142.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004334.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11004334.hg.1 |
TC11000342.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000529759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000529759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000534091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000534091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
ENST00000534091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151451.hg.1 |
TC1100007293.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151448.hg.1 |
TC1100007293.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000524419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000528989.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000529759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000529759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000532490.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000534091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000534091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
ENST00000534091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304263.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_001304264.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007293.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151453.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151453.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151453.hg.1 |
ENST00000315571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151453.hg.1 |
NM_174902.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151453.hg.1 |
TC1100007293.hg.1 |
Clariom D TC |