Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020106.hg.1 |
ENST00000305748.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020106.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020106.hg.1 |
TC12001530.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020110.hg.1 |
ENST00000305748.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020110.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020110.hg.1 |
TC12001530.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020108.hg.1 |
ENST00000305748.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020108.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020108.hg.1 |
TC12001530.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020105.hg.1 |
ENST00000305748.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020105.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020105.hg.1 |
TC12001530.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020107.hg.1 |
ENST00000305748.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020107.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020107.hg.1 |
TC12001530.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020109.hg.1 |
ENST00000305748.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020109.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12020109.hg.1 |
TC12001530.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182108.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182108.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182108.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182108.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182108.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182108.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182108.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182108.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182108.hg.1 |
TC1200010764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200182110.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182110.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182110.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182110.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182110.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182110.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182110.hg.1 |
TC1200010764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200182113.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182113.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182113.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182113.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182113.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182113.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182113.hg.1 |
TC1200010764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200182111.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182111.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182111.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182111.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182111.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182111.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182111.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182111.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182111.hg.1 |
TC1200010764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200182107.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182107.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182107.hg.1 |
TC1200010764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010764.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182109.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182109.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182109.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182109.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182109.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182109.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182109.hg.1 |
ENST00000552855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182109.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182109.hg.1 |
TC1200010764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200182114.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182114.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182114.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182114.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182114.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182114.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182114.hg.1 |
TC1200010764.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200182115.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182115.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182115.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182115.hg.1 |
ENST00000305748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182115.hg.1 |
ENST00000546934.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200182115.hg.1 |
NM_175068.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200182115.hg.1 |
TC1200010764.hg.1 |
Clariom D TC |