Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008379.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008379.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008379.hg.1 |
TC22000466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008382.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008382.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008382.hg.1 |
TC22000466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008381.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008381.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008381.hg.1 |
TC22000466.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155981.hg.1 |
TC2200007876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200155981.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155981.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155981.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155981.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155982.hg.1 |
TC2200007876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200155982.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155982.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155982.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155982.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007876.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155978.hg.1 |
TC2200007876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200155978.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155978.hg.1 |
NM_175878.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200155978.hg.1 |
ENST00000331428.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200155978.hg.1 |
NM_175878.4 |
Refseq |