Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022293.hg.1 |
TC05001641.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022293.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022293.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180267.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180267.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180267.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180267.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180267.hg.1 |
TC0500011596.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
ENST00000310954.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011596.hg.1 |
NM_180991.4 |
Refseq |