Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019075.hg.1 |
ENST00000304863.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019075.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019075.hg.1 |
TC19001379.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007412.hg.1 |
ENST00000435169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009321.hg.1 |
ENST00000426701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009321.hg.1 |
ENST00000426701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019072.hg.1 |
ENST00000304863.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019072.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19019072.hg.1 |
TC19001379.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900184515.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900184515.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900184515.hg.1 |
TC1900010286.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900184515.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900184515.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010286.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007412.hg.1 |
ENST00000435169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900184519.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900184519.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900184519.hg.1 |
TC1900010286.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900184519.hg.1 |
ENST00000304863.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900184519.hg.1 |
NM_006003.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009321.hg.1 |
ENST00000426701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009321.hg.1 |
ENST00000426701.1 |
Ensembl |