Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003475.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003475.hg.1 |
ENST00000376855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003475.hg.1 |
ENST00000376873.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003475.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003475.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003475.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003475.hg.1 |
TC13000335.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376855.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007752.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
ENST00000376855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
ENST00000299339.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
ENST00000376855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
ENST00000376873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
NM_001160100.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
NM_006984.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
NM_182848.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152235.hg.1 |
TC1300007752.hg.1 |
Clariom D TC |