Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003086.hg.1 |
ENST00000457354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003086.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14003086.hg.1 |
TC14000294.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400149459.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149459.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149459.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149459.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149459.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149459.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149459.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400149459.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400149459.hg.1 |
TC1400007139.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000457354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000555574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
ENST00000556683.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007139.hg.1 |
NM_030755.4 |
Refseq |