Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000506482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000506482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000506482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000506482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000506482.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000522676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000522676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000522676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000522676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
ENST00000522676.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019459.hg.1 |
TC04001399.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019459.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019459.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019460.hg.1 |
TC04001399.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019460.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04019460.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000506482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000506482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000506482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000506482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000506482.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000522676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000522676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000522676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000522676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
ENST00000522676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011374.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177746.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177746.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177746.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177746.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177746.hg.1 |
TC0400011374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400177747.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177747.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177747.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177747.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177747.hg.1 |
TC0400011374.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400177745.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177745.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177745.hg.1 |
ENST00000295268.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400177745.hg.1 |
NM_174952.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400177745.hg.1 |
TC0400011374.hg.1 |
Clariom D TC |