Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019802.hg.1 |
TC02001297.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019802.hg.1 |
ENST00000258362.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019802.hg.1 |
ENST00000273077.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019802.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019802.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02019802.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000469689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000469689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000469689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000469689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000469689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000472650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000472650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000472650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000483797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000483797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000483797.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000494954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000494954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000494954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
ENST00000622701.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010791.hg.1 |
NR_106868.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200176641.hg.1 |
TC0200010791.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000248451.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000258362.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000273077.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000436005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000469689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000469689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000469689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000469689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000469689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000472650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000472650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000472650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000483797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000483797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000483797.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000494954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000494954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000494954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
ENST00000622701.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_001077399.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_015488.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NM_022572.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010791.hg.1 |
NR_106868.1 |
Refseq |