Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
ENST00000274938.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_001303136.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007788.hg.1 |
NM_152753.3 |
Refseq |