Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009902.hg.1 |
ENST00000414124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000244020.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000483871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000483871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000483871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000483871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000483871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000483871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000483871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000483871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
ENST00000483871.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NM_006275.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007430.hg.1 |
NR_034009.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009902.hg.1 |
ENST00000414124.1 |
Ensembl |