Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
NM_194279_at |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7975711 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7975709 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7975707 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7975710 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7975708 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
7975712 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7975705 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
226007_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11720636_PM_s_at |
- |
- |
Affymetrix Human Genome U219 Array |
11720636_s_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005727.hg.1 |
ENST00000298818.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005727.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005727.hg.1 |
ENST00000556816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005727.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005727.hg.1 |
TC14000458.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005730.hg.1 |
ENST00000556816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005730.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005730.hg.1 |
TC14000458.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005724.hg.1 |
ENST00000298818.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005724.hg.1 |
ENST00000556816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005724.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005724.hg.1 |
TC14000458.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008309.hg.1 |
ENST00000425548.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005728.hg.1 |
ENST00000298818.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005728.hg.1 |
ENST00000556816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005728.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005728.hg.1 |
TC14000458.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005722.hg.1 |
ENST00000298818.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005722.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005722.hg.1 |
ENST00000556816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005722.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005722.hg.1 |
TC14000458.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005723.hg.1 |
ENST00000298818.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005723.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005723.hg.1 |
ENST00000556816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005723.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005723.hg.1 |
TC14000458.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005729.hg.1 |
ENST00000556816.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005729.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14005729.hg.1 |
TC14000458.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154190.hg.1 |
TC1400007679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154193.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154193.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154193.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154193.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154193.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154193.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154193.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154193.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154193.hg.1 |
TC1400007679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154192.hg.1 |
TC1400007679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154195.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154195.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154195.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154195.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154195.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154195.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154195.hg.1 |
TC1400007679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC2200008309.hg.1 |
ENST00000425548.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154183.hg.1 |
TC1400007679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154194.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154194.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154194.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154194.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154194.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154194.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154194.hg.1 |
TC1400007679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007679.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154185.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154185.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154185.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154185.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154185.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154185.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154185.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154185.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154185.hg.1 |
TC1400007679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
ENST00000298818.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
ENST00000554924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
ENST00000555139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154184.hg.1 |
TC1400007679.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400154178.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154178.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154178.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154178.hg.1 |
ENST00000556816.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400154178.hg.1 |
NM_001272007.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154178.hg.1 |
NM_194279.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400154178.hg.1 |
TC1400007679.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
226007_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
226007_at |
- |
- |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) |
NM_194279_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
226007_at5 |
- |
- |
Human Genome U95 Set |
53993_PM_at |
- |
- |
Human Genome U95 Set |
53993_at |
- |
- |
Human Genome U133 Set |
226007_at |
- |
- |
Human Genome U133 Set |
226007_PM_at |
- |
- |