Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026225.hg.1 |
ENST00000298966.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026225.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026225.hg.1 |
ENST00000527149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026225.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026225.hg.1 |
TC11002194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026223.hg.1 |
ENST00000298966.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026223.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026223.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026223.hg.1 |
TC11002194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026231.hg.1 |
ENST00000298966.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026231.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026231.hg.1 |
TC11002194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026226.hg.1 |
ENST00000298966.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026226.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026226.hg.1 |
ENST00000527149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026226.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026226.hg.1 |
TC11002194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026228.hg.1 |
ENST00000298966.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026228.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026228.hg.1 |
ENST00000527149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026228.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026228.hg.1 |
TC11002194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
ENST00000596676.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026224.hg.1 |
ENST00000298966.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026224.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026224.hg.1 |
ENST00000527149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026224.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026224.hg.1 |
TC11002194.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192261.hg.1 |
TC1100011973.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100192255.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192255.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192255.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192255.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192255.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192255.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192255.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192255.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192255.hg.1 |
TC1100011973.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
ENST00000596676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011973.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192254.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192254.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192254.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192254.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192254.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192254.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192254.hg.1 |
TC1100011973.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000596676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
ENST00000596676.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192258.hg.1 |
TC1100011973.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
ENST00000525141.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
ENST00000527149.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
ENST00000529714.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192259.hg.1 |
TC1100011973.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100192265.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192265.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192265.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192265.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192265.hg.1 |
ENST00000526869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192265.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192265.hg.1 |
TC1100011973.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100192266.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192266.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192266.hg.1 |
ENST00000298966.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100192266.hg.1 |
NM_020179.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100192266.hg.1 |
TC1100011973.hg.1 |
Clariom D TC |