Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286130.1 | NP_001273059.1 | 22 - 23 | 2262 | 143 | |
NM_001297.4 | NP_001288.3 | 22 - 23 | 2280 | 143 | ||
XM_011522870.2 | XP_011521172.1 | 10 - 11 | 1549 | 143 | ||
GenBank mRNA | AF042498.1 | - | 22 - 23 | 2267 | 143 | |
U58837.1 | - | 22 - 23 | 2272 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Homo sapiens Probe spans exons |