Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000529380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000529380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000529380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000529380.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000529897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000529897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000529897.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100173611.hg.1 |
TC1100009740.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000329957.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526462.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000526698.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000529380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000529380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000529380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000529380.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000529897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000529897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000529897.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000531976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
ENST00000614710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_001172223.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009740.hg.1 |
NM_053005.5 |
Refseq |