Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003361.hg.1 |
TC09000320.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003361.hg.1 |
ENST00000257515.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003361.hg.1 |
ENST00000303068.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003361.hg.1 |
ENST00000377225.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003361.hg.1 |
ENST00000455972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003361.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003361.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000469179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000469179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000469179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
ENST00000469179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900150866.hg.1 |
TC0900007505.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000257515.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000303068.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000377216.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000455972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000460871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000469179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000469179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000469179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
ENST00000469179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_001127608.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007505.hg.1 |
NM_004816.4 |
Refseq |