Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014078.hg.1 |
ENST00000328654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014078.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014078.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014078.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014078.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014078.hg.1 |
TC12001067.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSRUn_gl000223000025.hg.1 |
TCUn_gl000223000005.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSRUn_gl000223000025.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSRUn_gl000223000025.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSRUn_gl000223000025.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000545101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000545101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000545101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000545101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
ENST00000545101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000537956.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000545101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000545101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000545101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000545101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
ENST00000545101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012734.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202170.hg.1 |
TC1200012734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
ENST00000540238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
ENST00000544181.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202169.hg.1 |
TC1200012734.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200202171.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202171.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202171.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202171.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202171.hg.1 |
ENST00000534834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202171.hg.1 |
NM_001256279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202171.hg.1 |
NM_001256280.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202171.hg.1 |
NM_019591.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202171.hg.1 |
TC1200012734.hg.1 |
Clariom D TC |