Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025472.hg.1 |
ENST00000264552.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025472.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19025472.hg.1 |
TC19001872.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000589101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000589978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000589978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000589978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000589978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000589978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000592570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
ENST00000592570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197233.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197233.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197233.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900197233.hg.1 |
TC1900011470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900197233.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197233.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900197233.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000264552.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000587845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000589101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000589978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000589978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000589978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000589978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000589978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000592570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
ENST00000592570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011470.hg.1 |
NM_014501.2 |
Refseq |