Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011984.hg.1 |
TC08000919.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011984.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011984.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011985.hg.1 |
TC08000919.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011985.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08011985.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000474.hg.1 |
TC08000051.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000474.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08000474.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009463.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009463.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009463.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009463.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009463.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009463.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009463.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145006.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145006.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145006.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145006.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145006.hg.1 |
TC0800006624.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800164716.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800164716.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164716.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800164716.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164716.hg.1 |
TC0800009463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800164717.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800164717.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164717.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800164717.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164717.hg.1 |
TC0800009463.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800145004.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145004.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145004.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145004.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145004.hg.1 |
TC0800006624.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800145003.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145003.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145003.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800145003.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800145003.hg.1 |
TC0800006624.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
ENST00000302247.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800006624.hg.1 |
NM_004942.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009463.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009463.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009463.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009463.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009463.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009463.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009463.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164715.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800164715.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164715.hg.1 |
ENST00000318157.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800164715.hg.1 |
NM_001205266.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800164715.hg.1 |
TC0800009463.hg.1 |
Clariom D TC |