Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16591360 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046469 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612062 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612091 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612096 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046491 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612072 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046475 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046472 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046448 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046473 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612076 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612102 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612070 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046474 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046511 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046496 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046492 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046495 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046456 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046517 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612057 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046433 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612048 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612095 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612079 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046466 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612071 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046440 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046465 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046500 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046501 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046470 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046494 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612053 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046435 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612100 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612077 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16591373 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16591370 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612073 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P16591359 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612103 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046488 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P08612104 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046462 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046481 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P29046497 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
110523 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
6229 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
964444 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
170547 |
AF027866.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
170547 |
BX641156.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
170547 |
D88575.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
170547 |
NM_001040147.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
170547 |
NM_003784.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
960099 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
655818 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
708200 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
694179 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P167473 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_24_P167473 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
9538 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P111446 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P111446 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
53753 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P02701457 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P19680523 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P19680528 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P19680515 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P39815168 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P19680519 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P167473 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_19_P00800810 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_24_P167473 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29046517 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612104 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612095 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612099 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29046501 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612100 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16591373 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612096 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612102 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29046511 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612099 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612100 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612095 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29046517 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29046511 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612096 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P16591373 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P29046501 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612102 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P08612104 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_24_P167473 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P111446 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08612105 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08612098 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08612095 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08612096 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08612103 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P08612099 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16591375 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16591374 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P16591376 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P08612096 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P08612097 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P08612098 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P19680519 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P111446 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P111446 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_24_P167473 |
- |
- |
Custom Human Agilent Array 11k, Chromosome 6, Expression |
3357 |
- |
- |
Custom Human Agilent Array 11k, Chromosome 6, Expression |
1269 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
AB102653.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
AK289754.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
BC025843.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
BC126229.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
EF506611.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
M74387.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
M77640.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
NM_000425.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
NM_024003.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
X58776.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
167353 |
X59847.1 |
genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
54672 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19680531 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19680519 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P02701460 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P19680533 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P19482486 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P111446 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612105 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612053 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612085 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612073 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612080 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612099 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612103 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612087 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612069 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16591368 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612062 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16591361 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612092 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612076 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16591360 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612066 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16591376 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612096 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612139 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612083 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612078 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612095 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612050 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P16591373 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612059 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612064 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612089 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P08612094 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P19680519 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P111446 |
- |
- |