Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366578 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366638 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165885 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165836 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165834 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165882 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366606 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165837 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366573 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366626 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165879 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165896 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165874 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165878 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165860 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165898 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366651 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165850 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366579 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165849 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366581 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165846 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165854 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366574 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165891 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366650 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165835 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15873331 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15873290 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165842 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165845 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165881 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165890 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P25366602 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165897 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P05165888 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15873344 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
140126 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
148473 |
AF001846.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
148473 |
AF001847.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
148473 |
AF077031.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
148473 |
AF150732.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
148473 |
AK310570.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
148473 |
AK310698.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
148473 |
NM_012411.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
148473 |
NM_015967.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
325765 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
278253 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
26227 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
361946 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
326839 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
71851 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P857527 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_24_P857527 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
14379 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
30712 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_24_P857527 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_24_P857527 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873360 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165882 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165894 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165883 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366651 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873355 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366658 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873358 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165881 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366636 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165903 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165885 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366647 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366643 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165898 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366638 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165879 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165888 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873344 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366654 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366634 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165889 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165891 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165900 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165878 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366643 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165882 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165885 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366651 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165881 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366658 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873360 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165891 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366654 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165900 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366647 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366638 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165894 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165889 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165898 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873355 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165903 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165878 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366634 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P25366636 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873344 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165879 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165888 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15873358 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P05165883 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_24_P857527 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P01613185 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_24_P857527 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P01613185 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P15899863 |
- |
- |
Agilent Human 1B Microarray G4111A |
A_32_P303840 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15873345 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165836 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165838 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165846 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165882 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165874 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165854 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165885 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15873275 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165868 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165891 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165844 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15873337 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165853 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165866 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165845 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15873333 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165851 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165877 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165871 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165839 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165840 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165896 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165856 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165863 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165849 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165880 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165903 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165861 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165890 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165858 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165898 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15873289 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165860 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165888 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165842 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165876 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P05165883 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_18_P15899863 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P01613185 |
- |
- |