Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030545.hg.1 |
TC07001981.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030545.hg.1 |
ENST00000223140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030545.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030545.hg.1 |
ENST00000483238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030545.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012915.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193217.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193217.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193217.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193217.hg.1 |
ENST00000467773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193217.hg.1 |
ENST00000483238.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193217.hg.1 |
NM_001080413.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193217.hg.1 |
TC0700012915.hg.1 |
Clariom D TC |