Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030637.hg.1 |
TC02001922.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030637.hg.1 |
ENST00000356214.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030637.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030637.hg.1 |
ENST00000409784.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030637.hg.1 |
ENST00000409892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030637.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030638.hg.1 |
TC02001922.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030638.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030638.hg.1 |
ENST00000409784.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02030638.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000398529.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000398529.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000398529.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000398529.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000398529.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000494188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000494188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
ENST00000494188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000398529.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000398529.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000398529.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000398529.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000398529.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000494188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000494188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
ENST00000494188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200012854.hg.1 |
NM_015543.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193635.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193635.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193635.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193635.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193635.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193635.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193635.hg.1 |
TC0200012854.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200193634.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193634.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193634.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193634.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193634.hg.1 |
ENST00000409751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193634.hg.1 |
ENST00000409784.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193634.hg.1 |
ENST00000409892.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200193634.hg.1 |
NM_004161.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200193634.hg.1 |
TC0200012854.hg.1 |
Clariom D TC |