Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009483.hg.1 |
ENST00000408947.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009483.hg.1 |
ENST00000408947.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700009483.hg.1 |
NM_176882.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014328.hg.1 |
TC07000929.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014328.hg.1 |
ENST00000408947.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07014328.hg.1 |
NM_176882.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700009483.hg.1 |
ENST00000408947.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009483.hg.1 |
ENST00000408947.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700009483.hg.1 |
NM_176882.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167334.hg.1 |
ENST00000408947.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167334.hg.1 |
NM_176882.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167334.hg.1 |
ENST00000408947.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167334.hg.1 |
NM_176882.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700167334.hg.1 |
TC0700009483.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700167333.hg.1 |
ENST00000408947.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167333.hg.1 |
ENST00000408947.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700167333.hg.1 |
TC0700009483.hg.1 |
Clariom D TC |