Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174932.2 | NP_777592.1 | 12 - 13 | 1260 | 85 | |
XM_011530088.2 | XP_011528390.1 | 14 - 15 | 1440 | 85 | ||
XM_011530089.1 | XP_011528391.1 | 14 - 15 | 1397 | 85 | ||
XM_011530090.1 | XP_011528392.1 | 13 - 14 | 1298 | 85 | ||
XM_011530091.2 | XP_011528393.1 | 13 - 14 | 1284 | 85 | ||
XM_017028740.1 | XP_016884229.1 | 13 - 14 | 1269 | 85 | ||
GenBank mRNA | AF465766.1 | - | 12 - 13 | 1260 | 85 | |
AK301685.1 | - | 12 - 13 | 1166 | 85 | ||
BC131582.1 | - | 12 - 13 | 1175 | 40112599 | 85 | |
CU456409.1 | - | 3 - 4 | 308 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |