Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002343.hg.1 |
TC6_dbb_hap3000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002343.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002343.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002386.hg.1 |
TC6_qbl_hap6000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002386.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002386.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016976.hg.1 |
TC06001504.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016976.hg.1 |
ENST00000376437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016976.hg.1 |
ENST00000376442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016976.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016976.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002103.hg.1 |
TC6_mcf_hap5000117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002103.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002103.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002187.hg.1 |
TC6_ssto_hap7000119.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002187.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002187.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001933.hg.1 |
TC6_mann_hap4000116.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001933.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001933.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002414.hg.1 |
TC6_cox_hap2000138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002414.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002414.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001934.hg.1 |
TC6_mann_hap4000116.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001934.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001934.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002104.hg.1 |
TC6_mcf_hap5000117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002104.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002104.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016975.hg.1 |
TC06001504.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016975.hg.1 |
ENST00000376437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016975.hg.1 |
ENST00000376442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016975.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016975.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002413.hg.1 |
TC6_cox_hap2000138.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002413.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002413.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002344.hg.1 |
TC6_dbb_hap3000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002344.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002344.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002387.hg.1 |
TC6_qbl_hap6000128.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002387.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002387.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002188.hg.1 |
TC6_ssto_hap7000119.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002188.hg.1 |
NM_001164239.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002188.hg.1 |
NM_003587.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000415603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011375.hg.1 |
ENST00000480966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176667.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176667.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176667.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176667.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176667.hg.1 |
TC0600011375.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600176668.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176668.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176668.hg.1 |
ENST00000376437.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176668.hg.1 |
ENST00000376442.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176668.hg.1 |
TC0600011375.hg.1 |
Clariom D TC |