Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028298.hg.1 |
ENST00000334801.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028298.hg.1 |
ENST00000392849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028298.hg.1 |
ENST00000431085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028298.hg.1 |
ENST00000525300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028298.hg.1 |
ENST00000526143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028298.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028298.hg.1 |
TC11002355.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028299.hg.1 |
ENST00000334801.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028299.hg.1 |
ENST00000392849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028299.hg.1 |
ENST00000431085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028299.hg.1 |
ENST00000525300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028299.hg.1 |
ENST00000526143.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028299.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028299.hg.1 |
TC11002355.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100205530.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100205530.hg.1 |
ENST00000530293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100205530.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100205530.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100205530.hg.1 |
ENST00000530293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100205530.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100205530.hg.1 |
TC1100013230.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000527266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
ENST00000530293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013230.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100205529.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100205529.hg.1 |
ENST00000530293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100205529.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100205529.hg.1 |
ENST00000334801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100205529.hg.1 |
ENST00000530293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100205529.hg.1 |
NM_182557.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100205529.hg.1 |
TC1100013230.hg.1 |
Clariom D TC |