Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000613187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000613187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000613187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000616479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
ENST00000616479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000621.hg.1 |
ENST00000359061.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000621.hg.1 |
ENST00000441003.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000621.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000621.hg.1 |
TC11000039.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000613187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000613187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000613187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000616479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
ENST00000616479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006506.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145237.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100145237.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145237.hg.1 |
ENST00000361558.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100145237.hg.1 |
NM_002457.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100145237.hg.1 |
TC1100006506.hg.1 |
Clariom D TC |