Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014577.hg.1 |
ENST00000301292.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014577.hg.1 |
ENST00000301293.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014577.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19014577.hg.1 |
TC19001066.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000589889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000589889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
ENST00000589889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174741.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174741.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174741.hg.1 |
ENST00000589889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174741.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900174741.hg.1 |
TC1900009352.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900174741.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174741.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174741.hg.1 |
ENST00000589889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900174741.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586582.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000586965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000589889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000589889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
ENST00000589889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009352.hg.1 |
NM_032108.3 |
Refseq |