Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FASTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258461.1 | NP_001245390.1 | 2 - 3 | 620 | 83 | |
NM_006712.4 | NP_006703.1 | 2 - 3 | 620 | 83 | ||
XM_005249932.1 | XP_005249989.1 | 2 - 3 | 812 | 83 | ||
XM_005249933.3 | XP_005249990.1 | 2 - 3 | 1018 | 83 | ||
XM_011515761.2 | XP_011514063.1 | 2 - 3 | 620 | 83 | ||
XM_011515762.2 | XP_011514064.1 | 2 - 3 | 620 | 83 | ||
XM_017011704.1 | XP_016867193.1 | 2 - 3 | 812 | 83 | ||
GenBank mRNA | AB209783.1 | - | 2 - 3 | 521 | 83 | |
AK223132.1 | - | 2 - 3 | 530 | 83 | ||
AK292232.1 | - | 2 - 3 | 505 | 83 | ||
BC000377.2 | - | 2 - 3 | 532 | 2820338 | 83 | |
BC011770.2 | - | 2 - 3 | 531 | 3831196 | 83 | |
X86779.1 | - | 2 - 3 | 524 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |